The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026275.4 | NP_080551.1 | 3 - 4 | 408 | 78 |
GenBank mRNA | AJ240086.1 | - | 3 - 4 | 367 | 78 |
AK003550.1 | - | 3 - 4 | 428 | 78 | |
AK007517.1 | - | 3 - 4 | 542 | 78 | |
AK075703.1 | - | 3 - 4 | 418 | 78 | |
AK075714.1 | - | 3 - 4 | 409 | 78 | |
BC011112.1 | - | 3 - 4 | 416 | 78 |