The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027717.1 | NP_081993.1 | 3 - 4 | 355 | 101 |
GenBank mRNA | AK016963.1 | - | 3 - 4 | 490 | 101 |
BC113750.1 | - | 3 - 4 | 355 | 101 |