Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wapal
Entrez Gene ID
218914
Gene Name
wings apart-like homolog (Drosophila)
Gene Aliases
A530089A20Rik, BC037674, DIF-2, FOE, WAPL
Chromosome Location
Chr.14: 34671487 - 34747983 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004436.4 NP_001004436.2 12 - 13 3138 151
NM_001301330.1 NP_001288259.1 12 - 13 3120 151
GenBank mRNA AB167349.1 - 12 - 13 2944 151
AK028167.1 - 10 - 11 1242 151
AK129105.1 - 12 - 13 3138 151
BC037674.1 - 6 - 7 685 151
BC139434.1 - 12 - 13 2893 151
BC145625.1 - 12 - 13 2875 151

View Full Product Details