Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wapal
Entrez Gene ID
218914
Gene Name
wings apart-like homolog (Drosophila)
Gene Aliases
A530089A20Rik, BC037674, DIF-2, FOE, WAPL
Chromosome Location
Chr.14: 34671487 - 34747983 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001004436.4 NP_001004436.2 3 - 4 2002 76
GenBank mRNA AB167349.1 - 3 - 4 1808 76
AK041191.1 - 3 - 4 1736 76
AK129105.1 - 3 - 4 2002 76
BC139434.1 - 3 - 4 1757 76

View Full Product Details