Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bmpr1a
Entrez Gene ID
12166
Gene Name
bone morphogenetic protein receptor, type 1A
Gene Aliases
1110037I22Rik, ALK3, AU045487, BMPR-IA, Bmpr
Chromosome Location
Chr.14: 34411061 - 34503532 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009758.4 NP_033888.2 5 - 6 740 65
GenBank mRNA AK132051.1 - 5 - 6 753 65
AY365062.1 - 3 - 4 334 65
BC042611.1 - 4 - 5 530 65
D16250.1 - 3 - 4 475 65
U04672.1 - 3 - 4 344 65
Z23154.1 - 3 - 4 334 65

View Full Product Details