The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007589.3 | NP_001007590.2 | 3 - 4 | 950 | 109 |
GenBank mRNA | AK005836.1 | - | 3 - 4 | 110 | |
BC082305.1 | - | 3 - 4 | 3310 | 109 |