The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001294138.1 | NP_001281067.1 | 17 - 18 | 1741 | 66 |
NM_001294140.1 | NP_001281069.1 | 17 - 18 | 1675 | 66 | |
NM_008194.3 | NP_032220.1 | 16 - 17 | 1657 | 66 | |
NM_212444.2 | NP_997609.1 | 16 - 17 | 1657 | 66 | |
GenBank mRNA | AK044308.1 | - | 17 - 18 | 1513 | 66 |
AK088373.1 | - | 16 - 17 | 1476 | 66 | |
AK169424.1 | - | 17 - 18 | 1547 | 66 | |
AK169525.1 | - | 16 - 17 | 1658 | 66 | |
BC003767.1 | - | 16 - 17 | 1460 | 66 | |
CF740682.1 | - | 1 - 2 | 136 | 66 | |
U48403.1 | - | 16 - 17 | 1434 | 66 |