The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163020.1 | NP_001156492.1 | 4 - 5 | 716 | 78 |
GenBank mRNA | AK122569.1 | - | - | 339 | 78 |
BC141239.1 | - | 3 - 4 | 321 | 78 | |
CA556185.2 | - | 3 - 4 | 308 | 78 |