The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033531.3 | NP_001028703.2 | 6 - 7 | 1666 | 64 |
NM_001163020.1 | NP_001156492.1 | 7 - 8 | 1765 | 64 | |
GenBank mRNA | AK122569.1 | - | - | 1382 | 64 |
AK134944.1 | - | 6 - 7 | 1638 | 64 | |
BC141239.1 | - | 6 - 7 | 1370 | 64 | |
BC145566.1 | - | 5 - 6 | 1262 | 64 |