Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Letm2
Entrez Gene ID
270035
Gene Name
leucine zipper-EF-hand containing transmembrane protein 2
Gene Aliases
6030453H13, D030041N04Rik
Chromosome Location
Chr.8: 25578481 - 25597899 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173012.3 NP_766600.2 6 - 7 1375 68
GenBank mRNA AK031563.1 - 6 - 7 1228 68
AK163507.1 - 6 - 7 1229 68
BC050979.1 - 2 - 3 280 68
BC055685.1 - 6 - 7 1375 68
BE648035.1 - 1 - 2 162 68

View Full Product Details