The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178791.4 | NP_848906.1 | 1 - 2 | 208 | 85 |
GenBank mRNA | AK033244.1 | - | 1 - 2 | 164 | 85 |
AK040699.1 | - | 1 - 2 | 55 | 85 | |
AK053685.1 | - | 1 - 2 | 209 | 85 |