Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam208a
Entrez Gene ID
218850
Gene Name
family with sequence similarity 208, member A
Gene Aliases
4732418E12, 4933409E02Rik, D14Abb1e
Chromosome Location
Chr.14: 27426658 - 27483555 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114879.1 NP_001108351.1 23 - 24 4660 81
GenBank mRNA AK173099.1 - 3 - 4 395 81

View Full Product Details