The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114879.1 | NP_001108351.1 | 14 - 15 | 2417 | 134 |
NM_028945.1 | NP_083221.1 | 14 - 15 | 2417 | 134 | |
GenBank mRNA | AK028620.1 | - | 1 - 2 | 168 | 134 |
AK044563.1 | - | 14 - 15 | 2418 | 134 |