Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tpte
Entrez Gene ID
234129
Gene Name
transmembrane phosphatase with tensin homology
Gene Aliases
Pten2
Chromosome Location
Chr.8: 22283441 - 22371418 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_199257.2 NP_954866.2 13 - 14 1017 90
GenBank mRNA BC107329.2 - 13 - 14 812 90

View Full Product Details