The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001035239.2 | NP_001030316.1 | 16 - 17 | 2325 | 74 |
NM_001035240.2 | NP_001030317.1 | 17 - 18 | 2361 | 74 | |
NM_001035241.2 | NP_001030318.1 | 16 - 17 | 2331 | 74 | |
NM_001035242.1 | NP_001030319.1 | 16 - 17 | 2325 | 74 | |
NM_001035243.2 | NP_001030320.1 | 15 - 16 | 2295 | 74 | |
NM_001035244.1 | NP_001030321.1 | 16 - 17 | 2364 | 74 | |
NM_177341.4 | NP_796315.2 | 17 - 18 | 2428 | 74 | |
GenBank mRNA | AJ544532.1 | - | 16 - 17 | 2325 | 74 |
AJ544533.1 | - | 17 - 18 | 2361 | 74 | |
AJ544534.1 | - | 16 - 17 | 2331 | 74 | |
AJ544535.1 | - | 16 - 17 | 2325 | 74 | |
AJ544536.1 | - | 15 - 16 | 2295 | 74 | |
AK044454.1 | - | 17 - 18 | 2431 | 74 | |
AK051867.1 | - | 12 - 13 | 1494 | 74 | |
AK142504.1 | - | 17 - 18 | 2281 | 74 | |
AK173218.1 | - | 16 - 17 | 2396 | 74 | |
BC141400.1 | - | 17 - 18 | 2404 | 74 | |
BC141412.1 | - | 17 - 18 | 2404 | 74 | |
HM064407.1 | - | 16 - 17 | 2417 | 74 | |
HM064408.1 | - | 16 - 17 | 2417 | 74 | |
HM064409.1 | - | 17 - 18 | 2453 | 74 | |
HM064410.1 | - | 17 - 18 | 2492 | 74 | |
HM064411.1 | - | 16 - 17 | 2363 | 74 | |
JF706722.1 | - | 17 - 18 | 2424 | 74 | |
JX644970.1 | - | 16 - 17 | 2417 | 74 | |
JX644971.1 | - | 16 - 17 | 2417 | 74 | |
JX644972.1 | - | 17 - 18 | 2453 | 74 | |
JX644973.1 | - | 16 - 17 | 2423 | 74 | |
JX644974.1 | - | 15 - 16 | 2387 | 74 | |
JX644975.1 | - | 17 - 18 | 2492 | 74 | |
JX644976.1 | - | 16 - 17 | 2363 | 74 | |
JX644977.1 | - | 17 - 18 | 2399 | 74 | |
JX644978.1 | - | 16 - 17 | 2369 | 74 | |
JX644979.1 | - | 18 - 19 | 2528 | 74 | |
JX644980.1 | - | 17 - 18 | 2498 | 74 | |
JX644981.1 | - | 16 - 17 | 2462 | 74 | |
JX644982.1 | - | 17 - 18 | 2444 | 74 | |
JX644986.1 | - | 17 - 18 | 2453 | 74 |