Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem50b
Entrez Gene ID
77975
Gene Name
transmembrane protein 50B
Gene Aliases
AU015466, AU019872, B230114J08Rik
Chromosome Location
Chr.16: 91574508 - 91597680 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030018.3 NP_084294.1 6 - 7 576 84
GenBank mRNA AK020977.1 - 6 - 7 581 84
AK098154.1 - 6 - 7 540 84
AK133966.1 - 6 - 7 604 84
AK151796.1 - 6 - 7 606 84
AK152190.1 - 6 - 7 592 84
AK152767.1 - 6 - 7 592 84
AK155082.1 - 6 - 7 629 84
AK170684.1 - 6 - 7 571 84
AK172335.1 - 6 - 7 564 84
BC004841.1 - 6 - 7 579 84

View Full Product Details