Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uty
Entrez Gene ID
22290
Gene Name
ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome
Gene Aliases
Hydb, mKIAA4057
Chromosome Location
Chr.Y: 1085187 - 1245754 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009484.2 NP_033510.2 16 - 17 2448 141
GenBank mRNA AF057367.1 - 16 - 17 2428 141
AK142344.1 - 19 - 20 2586 141
AK142512.1 - 16 - 17 2364 141
AK162410.1 - 14 - 15 2021 141
AK164917.1 - 3 - 4 893 141
AK220463.1 - 17 - 18 2405 141
BC053061.1 - 16 - 17 2431 140

View Full Product Details