Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uty
Entrez Gene ID
22290
Gene Name
ubiquitously transcribed tetratricopeptide repeat gene, Y chromosome
Gene Aliases
Hydb, mKIAA4057
Chromosome Location
Chr.Y: 1085187 - 1245754 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009484.2 NP_033510.2 26 - 27 3799 90
GenBank mRNA AK142512.1 - 26 - 27 3715 90
AK162410.1 - 24 - 25 3372 90
AK164917.1 - 13 - 14 2244 90
AK220463.1 - 27 - 28 3756 90
BC053061.1 - 26 - 27 3781 90

View Full Product Details