The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012011.1 | NP_036141.1 | 4 - 5 | 387 | 101 |
GenBank mRNA | AJ006584.1 | - | 4 - 5 | 387 | 101 |
AK009448.1 | - | 4 - 5 | 392 | 101 | |
AK153974.1 | - | 4 - 5 | 405 | 101 | |
BC031450.1 | - | 4 - 5 | 383 | 101 | |
BC043656.1 | - | 4 - 5 | 387 | 101 |