The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019724.3 | NP_062698.2 | 8 - 9 | 1611 | 79 |
GenBank mRNA | AB021228.1 | - | 8 - 9 | 1426 | 79 |
AK048543.1 | - | 8 - 9 | 1491 | 79 | |
AK077939.1 | - | - | 1784 | 79 | |
AK163633.1 | - | 1 - 2 | 2167 | 79 | |
AK163638.1 | - | 1 - 2 | 2166 | 79 | |
BC057926.1 | - | 8 - 9 | 1613 | 79 |