Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Hadha
Entrez Gene ID
97212
Gene Name
hydroxyacyl-Coenzyme A dehydrogenase/3-ketoacyl-Coenzyme A thiolase/enoyl-Coenzyme A hydratase (trifunctional protein), alpha subunit
Gene Aliases
C77020, Mtpa
Chromosome Location
Chr.5: 30118304 - 30154980 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178878.2 NP_849209.1 12 - 13 1250 151
GenBank mRNA AK029017.1 - 12 - 13 1251 151
AK150188.1 - 12 - 13 1248 151
AK170478.1 - 12 - 13 1251 151
AK170683.1 - 12 - 13 1251 151
AK171469.1 - 12 - 13 1251 151
AK194527.1 - 1 - 2 139 151
BC027156.1 - 1 - 2 145 151
BC037009.1 - 12 - 13 1238 151
BC046978.1 - 12 - 13 1224 151
BC058569.1 - 12 - 13 1246 151

View Full Product Details