The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178878.2 | NP_849209.1 | 19 - 20 | 2180 | 65 |
GenBank mRNA | AK029017.1 | - | 19 - 20 | 2181 | 65 |
AK170683.1 | - | 19 - 20 | 2181 | 65 | |
BC027156.1 | - | 8 - 9 | 1075 | 65 | |
BC037009.1 | - | 19 - 20 | 2168 | 65 | |
BC046978.1 | - | 19 - 20 | 2154 | 65 | |
BC058569.1 | - | 19 - 20 | 2176 | 65 |