Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Htra4
Entrez Gene ID
330723
Gene Name
HtrA serine peptidase 4
Gene Aliases
B430206E18Rik
Chromosome Location
Chr.8: 25024928 - 25038962 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081187.3 NP_001074656.1 4 - 5 1073 104
GenBank mRNA AK046614.1 - 4 - 5 992 104
BC132380.1 - 4 - 5 1033 104
BC145842.1 - 4 - 5 1033 104

View Full Product Details