Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stk32b
Entrez Gene ID
64293
Gene Name
serine/threonine kinase 32B
Gene Aliases
2510009F08Rik, STKG6, Stk32, YANK2
Chromosome Location
Chr.5: 37446825 - 37717153 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_022416.2 NP_071861.1 7 - 8 1030 65
GenBank mRNA AJ250840.1 - 7 - 8 1108 65
AK082468.1 - 7 - 8 1031 65
BC052404.1 - 7 - 8 940 65
BC056396.1 - 7 - 8 1016 65
BC058412.1 - 7 - 8 940 65

View Full Product Details