The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024458.3 | NP_001019629.2 | 4 - 5 | 724 | 86 |
NM_001102444.1 | NP_001095914.1 | 4 - 5 | 724 | 86 | |
NM_013457.3 | NP_038485.1 | 4 - 5 | 724 | 86 | |
GenBank mRNA | AF096839.1 | - | 4 - 5 | 622 | 86 |
AF189771.1 | - | 3 - 4 | 504 | 86 | |
AK054558.1 | - | 4 - 5 | 576 | 86 | |
AK079960.1 | - | 4 - 5 | 579 | 86 | |
AK190114.1 | - | 1 - 2 | 69 | 86 | |
AK195469.1 | - | 1 - 2 | 69 | 86 | |
AK202054.1 | - | - | 218 | 86 | |
AK203448.1 | - | 1 - 2 | 69 | 86 | |
AK207989.1 | - | 1 - 2 | 69 | 86 | |
BC034368.1 | - | 3 - 4 | 555 | 86 |