The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024241.2 | NP_077203.2 | 6 - 7 | 1361 | 67 |
GenBank mRNA | AK050683.1 | - | 6 - 7 | 1347 | 67 |
AK135307.1 | - | 6 - 7 | 1362 | 67 | |
BC067395.1 | - | 6 - 7 | 1353 | 67 |