Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Srp72
Entrez Gene ID
66661
Gene Name
signal recognition particle 72
Gene Aliases
5730576P14Rik, 72kDa, AI132477, BC019196, C77589
Chromosome Location
Chr.5: 76974701 - 76999935 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025691.1 NP_079967.1 15 - 16 1532 91
GenBank mRNA AK017869.1 - 1 - 2 88 91
AK133784.1 - 15 - 16 1533 91
BC003434.1 - 15 - 16 1520 91
BC089462.1 - 6 - 7 540 91
BY393170.1 - 3 - 4 249 91

View Full Product Details