The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177561.3 | NP_808229.1 | 1 - 2 | 165 | 140 |
GenBank mRNA | AK145298.1 | - | 1 - 2 | 185 | 140 |
BC039916.1 | - | 1 - 2 | 167 | 140 |