Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dcun1d4
Entrez Gene ID
100737
Gene Name
DCN1, defective in cullin neddylation 1, domain containing 4 (S. cerevisiae)
Gene Aliases
AI836376
Chromosome Location
Chr.5: 73481027 - 73560794 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001190733.1 NP_001177662.1 6 - 7 555 70
NM_001190734.1 NP_001177663.1 7 - 8 490 70
NM_178896.5 NP_849227.1 6 - 7 555 70
GenBank mRNA AK033559.1 - 6 - 7 500 70
AK076991.1 - 7 - 8 491 70
BC060213.1 - 6 - 7 497 70
BC108346.1 - 6 - 7 457 70
BC137627.1 - 6 - 7 499 70
BC137628.1 - 6 - 7 499 70
BC144794.1 - 6 - 7 499 70
CO433728.1 - 2 - 3 164 70

View Full Product Details