The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_173765.3 | NP_776126.1 | 4 - 5 | 647 | 131 |
GenBank mRNA | AB095954.1 | - | 4 - 5 | 664 | 131 |
AK038779.1 | - | 5 - 6 | 657 | 131 | |
AK043807.1 | - | 4 - 5 | 625 | 131 | |
AK136849.1 | - | 4 - 5 | 645 | 131 | |
BC058102.1 | - | 3 - 4 | 559 | 131 | |
BC094507.1 | - | 3 - 4 | 424 | 131 | |
BC128330.1 | - | 3 - 4 | 519 | 131 |