Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map2k7
Entrez Gene ID
26400
Gene Name
mitogen-activated protein kinase kinase 7
Gene Aliases
5930412N11Rik, JNKK 2, Jnkk2, MAPKK 7, MEK 7, Mapkk7, Mek7, Mkk7, Prkmk7, sek2
Chromosome Location
Chr.8: 4238740 - 4247897 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042557.2 NP_001036022.1 10 - 11 1295 133
NM_001164172.1 NP_001157644.1 10 - 11 1295 133
NM_001291777.1 NP_001278706.1 9 - 10 1247 133
NM_001291778.1 NP_001278707.1 9 - 10 1138 133
NM_001291783.1 NP_001278712.1 9 - 10 1138 133
NM_011944.3 NP_036074.2 9 - 10 1247 133
GenBank mRNA AB005654.1 - 10 - 11 1170 133
AF060943.1 - 9 - 10 1138 136
AF060944.1 - 9 - 10 1158 133
AF060945.1 - 9 - 10 1158 136
AF060946.1 - 10 - 11 1206 133
AF060947.1 - 10 - 11 1206 136
AK031137.1 - 9 - 10 1156 133
AK165184.1 - 9 - 10 1164 133
AK219049.1 - 3 - 4 279 133
BC070467.1 - 10 - 11 1207 133
U74463.1 - 9 - 10 1104 133
U74464.1 - 8 - 9 993 133
U93030.1 - 9 - 10 1138 133

View Full Product Details