Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Map2k7
Entrez Gene ID
26400
Gene Name
mitogen-activated protein kinase kinase 7
Gene Aliases
5930412N11Rik, JNKK 2, Jnkk2, MAPKK 7, MEK 7, Mapkk7, Mek7, Mkk7, Prkmk7, sek2
Chromosome Location
Chr.8: 4238740 - 4247897 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042557.2 NP_001036022.1 2 - 3 342 77
NM_001164172.1 NP_001157644.1 2 - 3 342 77
GenBank mRNA AB005654.1 - 2 - 3 217 77
AF022113.1 - 2 - 3 319 77
AF060946.1 - 2 - 3 253 77
AF060947.1 - 2 - 3 253 77
BC070467.1 - 2 - 3 254 77

View Full Product Details