Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Map2k7
Entrez Gene ID
26400
Gene Name
mitogen-activated protein kinase kinase 7
Gene Aliases
5930412N11Rik, JNKK 2, Jnkk2, MAPKK 7, MEK 7, Mapkk7, Mek7, Mkk7, Prkmk7, sek2
Chromosome Location
Chr.8: 4238740 - 4247897 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001042557.2 NP_001036022.1 7 - 8 889 84
NM_001164172.1 NP_001157644.1 7 - 8 889 84
NM_001291777.1 NP_001278706.1 6 - 7 841 84
NM_001291778.1 NP_001278707.1 6 - 7 732 84
NM_001291783.1 NP_001278712.1 6 - 7 732 84
NM_011944.3 NP_036074.2 6 - 7 841 84
GenBank mRNA AB005654.1 - 7 - 8 764 84
AF022112.1 - 6 - 7 818 84
AF022113.1 - 7 - 8 866 84
AF060943.1 - 6 - 7 732 84
AF060944.1 - 6 - 7 752 84
AF060945.1 - 6 - 7 752 84
AF060946.1 - 7 - 8 800 84
AF060947.1 - 7 - 8 800 84
AK031137.1 - 6 - 7 750 84
AK165184.1 - 6 - 7 758 84
BC070467.1 - 7 - 8 801 84
U74463.1 - 6 - 7 698 84
U74464.1 - 5 - 6 587 84
U93030.1 - 6 - 7 732 84

View Full Product Details