The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012450.1 | NP_001012453.1 | 16 - 17 | 2126 | 56 |
NM_001012451.1 | NP_001012454.1 | 15 - 16 | 1837 | 56 | |
NM_080471.3 | NP_536719.2 | 15 - 16 | 1966 | 56 | |
GenBank mRNA | AK133551.1 | - | 14 - 15 | 1853 | 56 |
AK173070.1 | - | 15 - 16 | 1877 | 56 | |
AY026320.2 | - | 14 - 15 | 1758 | 56 | |
BC065177.1 | - | 16 - 17 | 2076 | 56 |