Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd6
Entrez Gene ID
140577
Gene Name
ankyrin repeat domain 6
Gene Aliases
-
Chromosome Location
Chr.4: 32804027 - 32950884 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012450.1 NP_001012453.1 9 - 10 1232 69
NM_001012451.1 NP_001012454.1 8 - 9 943 69
NM_080471.3 NP_536719.2 8 - 9 1072 69
GenBank mRNA AK133551.1 - 7 - 8 959 69
AK173070.1 - 8 - 9 983 69
AY026320.2 - 7 - 8 864 69
BC065177.1 - 9 - 10 1182 69

View Full Product Details