Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casp8ap2
Entrez Gene ID
26885
Gene Name
caspase 8 associated protein 2
Gene Aliases
AA387232, D4Ertd659e, FLASH, mKIAA1315
Chromosome Location
Chr.4: 32615473 - 32653271 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122978.1 NP_001116450.1 8 - 9 5785 112
NM_011997.2 NP_036127.2 8 - 9 6032 112
GenBank mRNA AF132726.1 - 8 - 9 5992 112
AK011818.1 - 1 - 2 443 112
BC138022.1 - 7 - 8 5775 112
BC138041.1 - 7 - 8 5775 112

View Full Product Details