The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001253743.1 | NP_001240672.1 | 1 - 2 | 91 | 76 |
NM_001253747.1 | NP_001240676.1 | 1 - 2 | 91 | 76 | |
NM_013517.4 | NP_038545.1 | 1 - 2 | 91 | 76 | |
GenBank mRNA | AK080159.1 | - | 1 - 2 | 92 | 76 |
AK156654.1 | - | 1 - 2 | 94 | 76 | |
AK172591.1 | - | 1 - 2 | 94 | 76 | |
M34163.1 | - | 1 - 2 | 93 | 76 | |
M99371.1 | - | 1 - 2 | 83 | 76 |