Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccl25
Entrez Gene ID
20300
Gene Name
chemokine (C-C motif) ligand 25
Gene Aliases
A130072A22Rik, AI852536, CKb15, Scya25, TECK
Chromosome Location
Chr.8: 4325159 - 4360020 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009138.3 NP_033164.1 5 - 6 526 88
NR_033527.1 5 - 6 551 88
GenBank mRNA AJ249480.1 - 5 - 6 499 88
AK132700.1 - 5 - 6 551 88
AK134138.1 - 5 - 6 511 88
BC117033.1 - 4 - 5 459 88
BC117037.1 - 4 - 5 459 88
DQ158256.1 - 5 - 6 512 88
U86357.1 - 4 - 5 408 88

View Full Product Details