The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007154.3 | NP_001007155.1 | 10 - 11 | 1441 | 133 |
NM_001177789.1 | NP_001171260.1 | 10 - 11 | 1659 | 133 | |
NM_001177790.1 | NP_001171261.1 | 10 - 11 | 1656 | 133 | |
NM_001177791.1 | NP_001171262.1 | 10 - 11 | 1493 | 133 | |
NM_028806.2 | NP_083082.1 | 10 - 11 | 1898 | 133 | |
GenBank mRNA | AJ632220.1 | - | 10 - 11 | 1456 | 133 |
AJ632221.1 | - | 10 - 11 | 1494 | 133 | |
AJ632222.1 | - | 10 - 11 | 1441 | 133 | |
AJ632223.1 | - | 9 - 10 | 1331 | 133 | |
AK005136.1 | - | 10 - 11 | 1463 | 133 | |
AK032242.1 | - | 10 - 11 | 1665 | 133 | |
AK038449.1 | - | 10 - 11 | 1409 | 133 | |
AK135144.1 | - | 10 - 11 | 1899 | 133 | |
AK220554.1 | - | 10 - 11 | 1656 | 133 | |
AY501387.1 | - | 10 - 11 | 1605 | 133 | |
BC058621.1 | - | 10 - 11 | 1814 | 133 | |
BC059869.1 | - | 10 - 11 | 1677 | 133 |