The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001007154.3 | NP_001007155.1 | 8 - 9 | 1318 | 62 |
NM_001177789.1 | NP_001171260.1 | 8 - 9 | 1536 | 62 | |
NM_001177791.1 | NP_001171262.1 | 8 - 9 | 1370 | 62 | |
NM_028806.2 | NP_083082.1 | 8 - 9 | 1775 | 62 | |
GenBank mRNA | AJ632220.1 | - | 8 - 9 | 1333 | 62 |
AJ632221.1 | - | 8 - 9 | 1371 | 62 | |
AJ632222.1 | - | 8 - 9 | 1318 | 62 | |
AJ632223.1 | - | 7 - 8 | 1208 | 62 | |
AK005136.1 | - | 8 - 9 | 1340 | 62 | |
AK032242.1 | - | 8 - 9 | 1542 | 62 | |
AK038449.1 | - | 8 - 9 | 1286 | 62 | |
AK039213.1 | - | 8 - 9 | 1325 | 62 | |
AK135144.1 | - | 8 - 9 | 1776 | 62 | |
AY501387.1 | - | 8 - 9 | 1482 | 62 | |
BC058621.1 | - | 8 - 9 | 1691 | 62 | |
BC059869.1 | - | 8 - 9 | 1554 | 62 |