The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164533.1 | NP_001158005.1 | 4 - 5 | 679 | 69 |
NM_022994.3 | NP_075370.2 | 4 - 5 | 679 | 69 | |
GenBank mRNA | AJ250375.1 | - | 4 - 5 | 423 | 69 |
AK014280.1 | - | 4 - 5 | 349 | 69 | |
AK014889.1 | - | 4 - 5 | 349 | 69 | |
AK032275.1 | - | 4 - 5 | 351 | 69 | |
AK088960.1 | - | 4 - 5 | 349 | 69 | |
BC019566.1 | - | 4 - 5 | 679 | 69 |