The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002927.2 | NP_001002927.1 | 1 - 2 | 447 | 57 |
GenBank mRNA | AK161272.1 | - | 2 - 3 | 303 | 57 |
BC049766.1 | - | 1 - 2 | 447 | 57 | |
M13227.1 | - | 1 - 2 | 61 | 57 |