Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gm41
Entrez Gene ID
245502
Gene Name
melanoma antigen, family B, 4 pseudogene
Gene Aliases
-
Chromosome Location
Chr.X: 86345475 - 86354994 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_036689.1 2 - 2 408 88
GenBank mRNA BB651730.1 - 2 - 2 409 88

View Full Product Details