The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030721.2 | NP_109646.2 | 18 - 19 | 2166 | 87 |
GenBank mRNA | AJ278430.1 | - | 18 - 19 | 2199 | 87 |
AK053960.1 | - | 18 - 19 | 2163 | 87 | |
AK128922.1 | - | 18 - 19 | 2126 | 87 | |
AK181413.1 | - | 3 - 4 | 213 | 87 | |
AK211732.1 | - | 3 - 4 | 213 | 87 | |
BC044725.2 | - | 18 - 19 | 2186 | 87 | |
BC055019.1 | - | 18 - 19 | 2166 | 87 |