The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
GenBank mRNA | AF099067.1 | - | 14 - 15 | 2424 | 71 |
AJ223071.1 | - | 14 - 15 | 2514 | 71 | |
AK078809.1 | - | 13 - 14 | 2391 | 71 | |
AK132803.1 | - | 13 - 14 | 2308 | 71 | |
AK147381.1 | - | 11 - 12 | 6211 | 71 | |
AK149147.1 | - | 5 - 6 | 592 | 71 | |
AK149584.1 | - | 13 - 14 | 2308 | 71 | |
BC057939.1 | - | 14 - 15 | 2426 | 71 | |
Y09234.1 | - | 13 - 14 | 2370 | 71 |