The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153547.6 | NP_705775.2 | 14 - 15 | 1726 | 78 |
NR_104298.1 | 14 - 15 | 1678 | 78 | ||
GenBank mRNA | AA041888.1 | - | - | 297 | 78 |
AK077523.1 | - | 14 - 15 | 1657 | 78 | |
AK167183.1 | - | 14 - 15 | 1610 | 78 | |
AY181025.1 | - | 14 - 15 | 1584 | 78 | |
AY185498.1 | - | 14 - 15 | 1485 | 78 | |
BC018560.1 | - | 14 - 15 | 1565 | 78 | |
BC037996.1 | - | 14 - 15 | 1586 | 78 |