The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_153547.6 | NP_705775.2 | 1 - 2 | 201 | 80 |
NR_104298.1 | 1 - 2 | 201 | 80 | ||
GenBank mRNA | AK077523.1 | - | 1 - 2 | 132 | 80 |
AK087757.1 | - | 1 - 2 | 85 | 80 | |
AK167183.1 | - | 1 - 2 | 85 | 80 | |
AY181025.1 | - | 1 - 2 | 59 | 80 | |
AY185498.1 | - | 1 - 2 | 59 | 80 | |
BC018560.1 | - | 1 - 2 | 88 | 80 | |
BC037996.1 | - | 1 - 2 | 61 | 80 | |
BY332148.1 | - | 1 - 2 | 160 | 80 |