The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001012517.5 | NP_001012535.1 | 1 - 2 | 130 | 145 |
NM_001286425.1 | NP_001273354.1 | 1 - 2 | 130 | 145 | |
NM_134161.3 | NP_598922.1 | 1 - 2 | 130 | 145 | |
GenBank mRNA | BC062113.1 | - | 1 - 2 | 129 | 145 |
BY721538.1 | - | 1 - 2 | 131 | 145 |