Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dthd1
Entrez Gene ID
100322896
Gene Name
death domain containing 1
Gene Aliases
Gm17384
Chromosome Location
Chr.5: 62813823 - 62888318 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001170705.1 NP_001164176.1 1 - 2 568 119

View Full Product Details