The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034862.2 | NP_001030034.2 | 5 - 6 | 987 | 91 |
GenBank mRNA | AK147828.1 | - | 5 - 6 | 963 | 91 |
AK170294.1 | - | 3 - 4 | 652 | 91 | |
CF981481.1 | - | 1 - 2 | 91 |